Product Details

SNP ID
rs147847190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144467493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCACCCTACTCCAGGATCCACA[C/T]CCCAAGAAGAAAGCCCTTCCCACTT
Phenotype
MIM: 616635 MIM: 603621 MIM: 615216
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYHR1 PubMed Links

Gene Details

Gene
CYHR1
Gene Name
cysteine and histidine rich 1
There are no transcripts associated with this gene.

Gene
FOXH1
Gene Name
forkhead box H1
There are no transcripts associated with this gene.

Gene
KIFC2
Gene Name
kinesin family member C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145754.3 561 Missense Mutation CCC,TCC P160S NP_665697.1
XM_005272357.3 561 Missense Mutation CCC,TCC P160S XP_005272414.1
XM_011517362.2 561 Missense Mutation CCC,TCC P160S XP_011515664.1

View Full Product Details