Product Details

SNP ID
rs148719420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:54137341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGATTTGGGTCGTGTTGATCCT[A/G]GTCAACCTATTGACTTAACCCAGCT
Phenotype
MIM: 611828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL15 PubMed Links

Gene Details

Gene
MRPL15
Gene Name
mitochondrial ribosomal protein L15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014175.3 411 Missense Mutation AGT,GGT S113G NP_054894.1
XM_017013362.1 411 Missense Mutation AGT,GGT S113G XP_016868851.1

View Full Product Details