Product Details

SNP ID
rs149279242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96273333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCAGAAGACAACATCTGGAGA[A/G]GCATCCTCTCTGTTATTTTCTTCTT
Phenotype
MIM: 612792
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTDSS1 PubMed Links

Gene Details

Gene
PTDSS1
Gene Name
phosphatidylserine synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290225.1 369 UTR 5 NP_001277154.1
NM_014754.2 369 Missense Mutation AGC,GGC S72G NP_055569.1

View Full Product Details