Product Details

SNP ID
rs149384065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22097229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGAGAACGGGCCCTCTGAGCC[A/G]GAACTGAGGGAGGAGTTGAGCCTGG
Phenotype
MIM: 615791
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM160B2 PubMed Links

Gene Details

Gene
FAM160B2
Gene Name
family with sequence similarity 160 member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022749.5 1621 Intron NP_073586.5
XM_005273602.3 1621 Intron XP_005273659.1
XM_006716381.2 1621 Intron XP_006716444.1
XM_006716385.3 1621 UTR 5 XP_006716448.1
XM_017013732.1 1621 Intron XP_016869221.1
XM_017013733.1 1621 Intron XP_016869222.1
XM_017013734.1 1621 Intron XP_016869223.1
Gene
NUDT18
Gene Name
nudix hydrolase 18
There are no transcripts associated with this gene.

View Full Product Details