Product Details

SNP ID
rs150510102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:490510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCGTCCCCATCTCCCCACACA[C/T]GTGATTTCCAGAGTTTCAAACACAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TDRP PubMed Links

Gene Details

Gene
TDRP
Gene Name
testis development related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256113.1 3017 UTR 3 NP_001243042.1
NM_175075.4 3017 UTR 3 NP_778250.2
XM_006716232.3 3017 UTR 3 XP_006716295.1
XM_017013113.1 3017 UTR 3 XP_016868602.1

View Full Product Details