Product Details

SNP ID
rs150751334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28770000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCCGCTTTCTCTTCTTCCCGCT[C/T]GTGGGCTGGGCGGGCCGAGGAGGGG
Phenotype
MIM: 611352
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INTS9 PubMed Links

Gene Details

Gene
INTS9
Gene Name
integrator complex subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145159.2 1785 Silent Mutation ACA,ACG T542T NP_001138631.1
NM_001172562.1 1785 Silent Mutation ACA,ACG T539T NP_001166033.1
NM_018250.3 1785 Silent Mutation ACA,ACG T563T NP_060720.2
XM_006716356.2 1785 Silent Mutation ACA,ACG T563T XP_006716419.1
XM_006716357.3 1785 Silent Mutation ACA,ACG T563T XP_006716420.1
XM_011544573.1 1785 Silent Mutation ACA,ACG T542T XP_011542875.1
XM_011544574.1 1785 Silent Mutation ACA,ACG T539T XP_011542876.1
XM_011544575.2 1785 Silent Mutation ACA,ACG T539T XP_011542877.1
XM_011544576.1 1785 Silent Mutation ACA,ACG T518T XP_011542878.1

View Full Product Details