Product Details

SNP ID
rs151071783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:107249874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGAACACAAAAGGACAAAATAC[A/T]CATTTTCTTTCCCTTAAATGAAAAC
Phenotype
MIM: 601667
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANGPT1 PubMed Links

Gene Details

Gene
ANGPT1
Gene Name
angiopoietin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146.4 3157 UTR 3 NP_001137.2
NM_001199859.2 3157 UTR 3 NP_001186788.1
NM_001314051.1 3157 UTR 3 NP_001300980.1

View Full Product Details