Product Details

SNP ID
rs138079601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92952969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGACTAAGGATTGAATAGTAAA[C/T]GGTGGATCCAAGTGATTGTGAGAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGD3 PubMed Links

Gene Details

Gene
FGD3
Gene Name
FYVE, RhoGEF and PH domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083536.1 Intron NP_001077005.1
NM_001286993.1 Intron NP_001273922.1
NM_033086.2 Intron NP_149077.2
XM_017015274.1 Intron XP_016870763.1

View Full Product Details