Product Details

SNP ID
rs138669342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97998562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCATTTTCTCTTGCAGCTTGAA[C/T]TCAGTGAAAATAGAATCTTTGGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANP32B PubMed Links

Gene Details

Gene
ANP32B
Gene Name
acidic nuclear phosphoprotein 32 family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006401.2 560 Missense Mutation CTC,TTC L71F NP_006392.1

View Full Product Details