Product Details

SNP ID
rs138679112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21202159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGACCATCAGTAAAGAAAAGGACC[A/G]GGCCATTGGGATTTTGCAAATATCA
Phenotype
MIM: 147577 MIM: 147567
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNA10 PubMed Links

Gene Details

Gene
IFNA10
Gene Name
interferon alpha 10
There are no transcripts associated with this gene.

Gene
IFNA7
Gene Name
interferon alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021057.2 47 Silent Mutation NP_066401.2

View Full Product Details