Product Details

SNP ID
rs138726067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:66918590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAGCCAGAGCTCAGCCCATATA[G/C]TACATCAGAAAACACAAGCTGGAGA
Phenotype
MIM: 616290
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF658 PubMed Links

Gene Details

Gene
ZNF658
Gene Name
zinc finger protein 658
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317916.1 Intron NP_001304845.1
NM_033160.6 Intron NP_149350.3
XM_005272515.4 Intron XP_005272572.1
XM_011545679.2 Intron XP_011543981.1
XM_017014614.1 Intron XP_016870103.1

View Full Product Details