Product Details

SNP ID
rs139680840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83625115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAACAATGCAGGTAAAGAGAAT[A/G]TGGTCATATGCTGGCAATATTTTGA
Phenotype
MIM: 611343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDNK PubMed Links

Gene Details

Gene
IDNK
Gene Name
IDNK, gluconokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001551.3 358 Intron NP_001001551.2
NM_001256915.1 358 Intron NP_001243844.1
XM_006717110.3 358 Intron XP_006717173.1
XM_006717111.3 358 UTR 5 XP_006717174.1
XM_017014719.1 358 Intron XP_016870208.1
XM_017014720.1 358 Intron XP_016870209.1
XM_017014721.1 358 Intron XP_016870210.1
XM_017014722.1 358 Intron XP_016870211.1
XM_017014723.1 358 Intron XP_016870212.1
XM_017014724.1 358 Intron XP_016870213.1
XM_017014725.1 358 Intron XP_016870214.1

View Full Product Details