Product Details

SNP ID
rs139732329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32630768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGTCTTAGTGTACTGACTCTC[A/G]GGTCCATTATACTTAACACTGTTGG
Phenotype
MIM: 607798
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAF1L PubMed Links

Gene Details

Gene
TAF1L
Gene Name
TATA-box binding protein associated factor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153809.2 4902 Silent Mutation CCC,CCT P1604P NP_722516.1

View Full Product Details