Product Details

SNP ID
rs140572425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127447877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAACAATGAAAATGTCACTTAC[A/G]GCTGGGCATTCCACAGCACCACTGT
Phenotype
MIM: 610933 MIM: 180475 MIM: 194552
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRSAM1 PubMed Links

Gene Details

Gene
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
There are no transcripts associated with this gene.

Gene
RPL12
Gene Name
ribosomal protein L12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000976.3 Intron NP_000967.1
Gene
SNORA65
Gene Name
small nucleolar RNA, H/ACA box 65
There are no transcripts associated with this gene.

Gene
ZNF79
Gene Name
zinc finger protein 79
There are no transcripts associated with this gene.

View Full Product Details