Product Details

SNP ID
rs140758818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:91409478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAGAAACTTTGTAGCCACTGTC[C/T]TTCATTTCAACAACTCCAGTTTTGA
Phenotype
MIM: 605327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFIL3 PubMed Links

Gene Details

Gene
NFIL3
Gene Name
nuclear factor, interleukin 3 regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289999.1 1790 Silent Mutation AAA,AAG K419K NP_001276928.1
NM_001290000.1 1790 Silent Mutation AAA,AAG K419K NP_001276929.1
NM_005384.2 1790 Silent Mutation AAA,AAG K419K NP_005375.2
XM_017014743.1 1790 Silent Mutation AAA,AAG K419K XP_016870232.1
XM_017014744.1 1790 Silent Mutation AAA,AAG K419K XP_016870233.1

View Full Product Details