Product Details

SNP ID
rs140834278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114589346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTCTATCCCTATTTTACAGAT[A/G]GTAAAAATGAGGCTCAGAGAGAAAT
Phenotype
MIM: 607296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1G1 PubMed Links

Gene Details

Gene
ATP6V1G1
Gene Name
ATPase H+ transporting V1 subunit G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004888.3 Intron NP_004879.1

View Full Product Details