Product Details

SNP ID
rs140946306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113386919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGCCAGATTCATCCTTCACAT[C/T]GAATCTACCTTCTAAACACATCCAC
Phenotype
MIM: 125270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALAD PubMed Links

Gene Details

Gene
ALAD
Gene Name
aminolevulinate dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000031.5 2742 UTR 3 NP_000022.3
NM_001003945.2 2742 UTR 3 NP_001003945.1
NM_001317745.1 2742 UTR 3 NP_001304674.1
XM_011518364.2 2742 UTR 3 XP_011516666.1
Gene
HDHD3
Gene Name
haloacid dehalogenase like hydrolase domain containing 3
There are no transcripts associated with this gene.

View Full Product Details