Product Details
- SNP ID
-
rs141926742
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:32784601 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCAGCTCCATCAACAGCCCCACA[A/C]CCACGGAGGAAGGAGAATGCCAGAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM215
PubMed Links
Gene Details
- Gene
- TMEM215
- Gene Name
- transmembrane protein 215
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_212558.2 |
783 |
Missense Mutation |
ACC,CCC |
T140P |
NP_997723.2 |
View Full Product Details