Product Details

SNP ID
rs141998405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122241887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTACAGTAATTCACAACCATCA[C/T]CTTCGAGATTTCCATGCTGTTCTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM18 PubMed Links

Gene Details

Gene
RBM18
Gene Name
RNA binding motif protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033117.3 736 Silent Mutation AGA,AGG R190R NP_149108.1
XM_006717319.1 736 Silent Mutation AGA,AGG R190R XP_006717382.1

View Full Product Details