Product Details

SNP ID
rs142724913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86292559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATGACAGAGGAAACGATGTTG[A/G]TGATGGTACTCAAGGCACTTCCACT
Phenotype
MIM: 611006 MIM: 613467
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISCA1 PubMed Links

Gene Details

Gene
ISCA1
Gene Name
iron-sulfur cluster assembly 1
There are no transcripts associated with this gene.

Gene
ZCCHC6
Gene Name
zinc finger CCHC-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185059.1 Intron NP_001171988.1
NM_001185074.1 Intron NP_001172003.1
NM_024617.3 Intron NP_078893.2
XM_005252207.3 Intron XP_005252264.1
XM_005252208.2 Intron XP_005252265.1
XM_006717283.1 Intron XP_006717346.1
XM_011519011.2 Intron XP_011517313.1
XM_011519012.2 Intron XP_011517314.1
XM_011519013.2 Intron XP_011517315.1
XM_011519014.2 Intron XP_011517316.1
XM_011519015.1 Intron XP_011517317.1
XM_011519016.1 Intron XP_011517318.1
XM_017015130.1 Intron XP_016870619.1
XM_017015131.1 Intron XP_016870620.1
XM_017015132.1 Intron XP_016870621.1

View Full Product Details