Product Details

SNP ID
rs143296733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96818594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCGCATTCATTACATTCAAAAG[A/G]TTTCTCCCCTGTGTGAGTTCTCTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF782 PubMed Links

Gene Details

Gene
ZNF782
Gene Name
zinc finger protein 782
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001662.1 1908 Missense Mutation CCT,TCT P477S NP_001001662.1
XM_005251742.4 1908 Missense Mutation CCT,TCT P477S XP_005251799.1
XM_005251744.3 1908 Missense Mutation CCT,TCT P453S XP_005251801.1
XM_011518315.2 1908 Missense Mutation CCT,TCT P477S XP_011516617.1
XM_011518318.2 1908 Missense Mutation CCT,TCT P477S XP_011516620.1
XM_011518320.2 1908 Missense Mutation CCT,TCT P471S XP_011516622.1
XM_017014343.1 1908 Missense Mutation CCT,TCT P471S XP_016869832.1
XM_017014344.1 1908 Missense Mutation CCT,TCT P345S XP_016869833.1

View Full Product Details