Product Details

SNP ID
rs143390652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19057967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCTATTTAAACCTTACCCTTTA[A/T]ATTAGGTAGTTCTCCAAGACTACAT
Phenotype
MIM: 613433 MIM: 612194 MIM: 616292 MIM: 615646
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HAUS6 PubMed Links

Gene Details

Gene
HAUS6
Gene Name
HAUS augmin like complex subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270890.1 1939 Intron NP_001257819.1
NM_017645.4 1939 Intron NP_060115.3
XM_011517935.2 1939 Missense Mutation ATA,TTA I503L XP_011516237.1
XM_017014830.1 1939 Intron XP_016870319.1
Gene
RRAGA
Gene Name
Ras related GTP binding A
There are no transcripts associated with this gene.

Gene
SAXO1
Gene Name
stabilizer of axonemal microtubules 1
There are no transcripts associated with this gene.

Gene
SCARNA8
Gene Name
small Cajal body-specific RNA 8
There are no transcripts associated with this gene.

View Full Product Details