Product Details

SNP ID
rs143679153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133360360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGAGGATGAGAAGGCAAAGCCC[C/T]CAAGAGAGAAGGCCACTGATGAGGG
Phenotype
MIM: 185640 MIM: 185620 MIM: 185630 MIM: 185660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL7A PubMed Links

Gene Details

Gene
RPL7A
Gene Name
ribosomal protein L7a
There are no transcripts associated with this gene.

Gene
SNORD36A
Gene Name
small nucleolar RNA, C/D box 36A
There are no transcripts associated with this gene.

Gene
SNORD36C
Gene Name
small nucleolar RNA, C/D box 36C
There are no transcripts associated with this gene.

Gene
SURF1
Gene Name
surfeit 1
There are no transcripts associated with this gene.

Gene
SURF2
Gene Name
surfeit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278928.1 661 Missense Mutation CCA,TCA P205S NP_001265857.1
NM_017503.4 661 Missense Mutation CCA,TCA P205S NP_059973.4
Gene
SURF4
Gene Name
surfeit 4
There are no transcripts associated with this gene.

View Full Product Details