Product Details

SNP ID
rs144419396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114092628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGGGGGCCTGCCACCTGAGGC[C/T]TCAGGGTCCAACACCTGTAGGAAAG
Phenotype
MIM: 611278
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF12 PubMed Links

Gene Details

Gene
KIF12
Gene Name
kinesin family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138424.1 1676 Silent Mutation GAA,GAG E399E NP_612433.1
XM_005251683.4 1676 Silent Mutation GAA,GAG E537E XP_005251740.1
XM_006716947.2 1676 Silent Mutation GAA,GAG E503E XP_006717010.1
XM_011518190.2 1676 Silent Mutation GAA,GAG E595E XP_011516492.1
XM_011518191.2 1676 Silent Mutation GAA,GAG E595E XP_011516493.1
XM_011518192.2 1676 UTR 3 XP_011516494.1
XM_011518193.2 1676 Intron XP_011516495.1
XM_017014240.1 1676 Silent Mutation GAA,GAG E272E XP_016869729.1

View Full Product Details