Product Details

SNP ID
rs145906216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130693857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCGAGAGACTGGGCCGCGACAC[C/T]AAGAAACATCTAGTGGTGCCGGGGG
Phenotype
MIM: 602238
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC2 PubMed Links

Gene Details

Gene
EXOSC2
Gene Name
exosome component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282708.1 98 Silent Mutation ACC,ACT T22T NP_001269637.1
NM_001282709.1 98 Silent Mutation ACC,ACT T22T NP_001269638.1
NM_014285.6 98 Silent Mutation ACC,ACT T22T NP_055100.2
XM_005272176.2 98 UTR 5 XP_005272233.1
XM_006717023.2 98 UTR 5 XP_006717086.1
XM_017014558.1 98 UTR 5 XP_016870047.1

View Full Product Details