Product Details

SNP ID
rs147602151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114093264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCTCCCCTGGCAGGCAGGCCC[A/C]GTGGGCCAGGGGCACTCGACACAGT
Phenotype
MIM: 611278
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIF12 PubMed Links

Gene Details

Gene
KIF12
Gene Name
kinesin family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138424.1 1626 Missense Mutation GGG,TGG G383W NP_612433.1
XM_005251683.4 1626 Missense Mutation GGG,TGG G521W XP_005251740.1
XM_006716947.2 1626 Missense Mutation GGG,TGG G487W XP_006717010.1
XM_011518190.2 1626 Missense Mutation GGG,TGG G579W XP_011516492.1
XM_011518191.2 1626 Missense Mutation GGG,TGG G579W XP_011516493.1
XM_011518192.2 1626 Missense Mutation GGG,TGG G579W XP_011516494.1
XM_011518193.2 1626 Intron XP_011516495.1
XM_017014240.1 1626 Missense Mutation GGG,TGG G256W XP_016869729.1

View Full Product Details