Product Details

SNP ID
rs147624133
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133349964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACAGACCAGAGACAAAGCAAGA[A/G]AAGAAGCAGAGACTGTTGGCCCGGG
Phenotype
MIM: 185641 MIM: 185640 MIM: 185620 MIM: 185630
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MED22 PubMed Links
Additional Information
For this assay, SNP(s) [rs7700] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MED22
Gene Name
mediator complex subunit 22
There are no transcripts associated with this gene.

Gene
RPL7A
Gene Name
ribosomal protein L7a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000972.2 Intron NP_000963.1
Gene
SNORD24
Gene Name
small nucleolar RNA, C/D box 24
There are no transcripts associated with this gene.

Gene
SNORD36A
Gene Name
small nucleolar RNA, C/D box 36A
There are no transcripts associated with this gene.

Gene
SNORD36B
Gene Name
small nucleolar RNA, C/D box 36B
There are no transcripts associated with this gene.

Gene
SNORD36C
Gene Name
small nucleolar RNA, C/D box 36C
There are no transcripts associated with this gene.

Gene
SURF1
Gene Name
surfeit 1
There are no transcripts associated with this gene.

Gene
SURF2
Gene Name
surfeit 2
There are no transcripts associated with this gene.

View Full Product Details