Product Details

SNP ID
rs148006496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122240395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGATTAACCTTGGGAAAAGTA[A/T]AAAGAATTTCCTTAAACCAGTTTTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RBM18 PubMed Links

Gene Details

Gene
RBM18
Gene Name
RNA binding motif protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033117.3 2228 UTR 3 NP_149108.1
XM_006717319.1 2228 UTR 3 XP_006717382.1

View Full Product Details