Product Details
- SNP ID
-
rs148296950
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:87883168 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACGTCCACAGTGACCCACCCTCA[A/C]CCCCGCCCGGGAGGAAGAGGAGCAG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC497256
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4072357] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC497256
- Gene Name
- uncharacterized LOC497256
There are no transcripts associated with this gene.
- Gene
- SPATA31E1
- Gene Name
- SPATA31 subfamily E member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178828.4 |
|
Intron |
|
|
NP_849150.3 |
View Full Product Details