Product Details

SNP ID
rs149375049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136755463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCCGCCACATCAGGGACACCC[A/G]CAGGATGGCGTAGCCCTCGTAGTCG
Phenotype
MIM: 609379 MIM: 612902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCN15 PubMed Links

Gene Details

Gene
LCN15
Gene Name
lipocalin 15
There are no transcripts associated with this gene.

Gene
LCN6
Gene Name
lipocalin 6
There are no transcripts associated with this gene.

Gene
LCN8
Gene Name
lipocalin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178469.3 629 Missense Mutation CGG,TGG R94W NP_848564.2
XM_017014272.1 629 Missense Mutation CGG,TGG R117W XP_016869761.1
Gene
LOC100128593
Gene Name
uncharacterized LOC100128593
There are no transcripts associated with this gene.

Gene
MIR6722
Gene Name
microRNA 6722
There are no transcripts associated with this gene.

View Full Product Details