Product Details

SNP ID
rs149588398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97309517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATGATGAAGAAGGTGGAGACTC[C/G]TGAAGGGGAGGTGATGTCTCCCCGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC180 PubMed Links

Gene Details

Gene
CCDC180
Gene Name
coiled-coil domain containing 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020893.2 433 Missense Mutation CCT,CGT P102R NP_065944.2
Gene
LOC100499484-C9ORF174
Gene Name
LOC100499484-C9orf174 readthrough
There are no transcripts associated with this gene.

View Full Product Details