Product Details
- SNP ID
-
rs149925472
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:35870043 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCAGATGGCCAGGTAGCGGTCA[C/T]AGGCCGTGATGGCCAGTAGCAGGCA
- Phenotype
-
MIM: 616888
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00950
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7044405] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC00950
- Gene Name
- long intergenic non-protein coding RNA 950
There are no transcripts associated with this gene.
- Gene
- OR13J1
- Gene Name
- olfactory receptor family 13 subfamily J member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004487.1 |
359 |
Missense Mutation |
TAT,TGT |
Y120C |
NP_001004487.1 |
- Gene
- TMEM8B
- Gene Name
- transmembrane protein 8B
There are no transcripts associated with this gene.
View Full Product Details