Product Details

SNP ID
rs150116611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35906547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCTCATGTGCCGAATGTGGGCC[A/T]CCACCACCACCACCACCCCCGCCAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HRCT1 PubMed Links

Gene Details

Gene
HRCT1
Gene Name
histidine rich carboxyl terminus 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039792.1 356 Missense Mutation CAC,CTC H87L NP_001034881.1
Gene
LINC00961
Gene Name
long intergenic non-protein coding RNA 961
There are no transcripts associated with this gene.

View Full Product Details