Product Details

SNP ID
rs150262376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:27110509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAAATACATTTATCATTGCCAT[C/T]AGTCTCTCCCCTTCGGTTTGAAAAC
Phenotype
MIM: 600221
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEK PubMed Links

Gene Details

Gene
TEK
Gene Name
TEK receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000459.4 Intron NP_000450.2
NM_001290077.1 Intron NP_001277006.1
NM_001290078.1 Intron NP_001277007.1
XM_005251561.1 Intron XP_005251618.1
XM_005251563.1 Intron XP_005251620.1

View Full Product Details