Product Details

SNP ID
rs150460415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128634735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAGTAGATGGGACCGCCGTGGG[G/T]GGAGAAGGTAAACTGTGCTGGGGCC
Phenotype
MIM: 182810 MIM: 613363
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPTAN1 PubMed Links

Gene Details

Gene
SPTAN1
Gene Name
spectrin alpha, non-erythrocytic 1
There are no transcripts associated with this gene.

Gene
WDR34
Gene Name
WD repeat domain 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052844.3 1292 Missense Mutation ACC,CCC T390P NP_443076.2
XM_011519179.2 1292 Missense Mutation ACC,CCC T362P XP_011517481.1

View Full Product Details