Product Details

SNP ID
rs150854491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97308109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCAGGTTCCGAATGGGAAAGCC[A/T]ACCAGCAGATCTTCCAGGCTGAGGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC180 PubMed Links

Gene Details

Gene
CCDC180
Gene Name
coiled-coil domain containing 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020893.2 306 Missense Mutation AAC,TAC N60Y NP_065944.2
Gene
LOC100499484-C9ORF174
Gene Name
LOC100499484-C9orf174 readthrough
There are no transcripts associated with this gene.

View Full Product Details