Product Details

SNP ID
rs113047476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154012670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGACGATGATGCAGAGCTGCCAA[A/G]GGCCAGTCCTGGGGTGGAGATGGCA
Phenotype
MIM: 300283 MIM: 300005 MIM: 300929
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRAK1 PubMed Links

Gene Details

Gene
IRAK1
Gene Name
interleukin 1 receptor associated kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025242.1 1950 Missense Mutation CTT,TTT L617F NP_001020413.1
NM_001025243.1 1950 Missense Mutation CTT,TTT L568F NP_001020414.1
NM_001569.3 1950 Missense Mutation CTT,TTT L647F NP_001560.2
XM_005274668.3 1950 Missense Mutation CTT,TTT L643F XP_005274725.1
XM_011531158.2 1950 Missense Mutation CTT,TTT L538F XP_011529460.1
Gene
MECP2
Gene Name
methyl-CpG binding protein 2
There are no transcripts associated with this gene.

Gene
MIR718
Gene Name
microRNA 718
There are no transcripts associated with this gene.

View Full Product Details