Product Details

SNP ID
rs137883992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155027046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGTGTGTCCTCGCGAGACAAGCT[C/T]ACCGAAATGGCCGCGTCCAGTCAAG
Phenotype
MIM: 300841
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F8 PubMed Links

Gene Details

Gene
F8
Gene Name
coagulation factor VIII
There are no transcripts associated with this gene.

Gene
FUNDC2
Gene Name
FUN14 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023934.3 258 Silent Mutation CTC,CTT L36L NP_076423.2

View Full Product Details