Product Details

SNP ID
rs137939858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102714932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGAAAAGAAGGCTGGGGAAGA[C/G]GTTATCGCTGGGCCTGAGAGAGAGA
Phenotype
MIM: 300921
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARMCX5-GPRASP2 PubMed Links

Gene Details

Gene
ARMCX5-GPRASP2
Gene Name
ARMCX5-GPRASP2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199818.1 1139 Missense Mutation GAC,GAG D21E NP_001186747.1
Gene
BHLHB9
Gene Name
basic helix-loop-helix domain containing, class B, 9
There are no transcripts associated with this gene.

Gene
GPRASP2
Gene Name
G protein-coupled receptor associated sorting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004051.3 1139 Missense Mutation GAC,GAG D21E NP_001004051.1
NM_001184874.2 1139 Missense Mutation GAC,GAG D21E NP_001171803.1
NM_001184875.2 1139 Missense Mutation GAC,GAG D21E NP_001171804.1
NM_001184876.2 1139 Missense Mutation GAC,GAG D21E NP_001171805.1
NM_138437.5 1139 Missense Mutation GAC,GAG D21E NP_612446.1

View Full Product Details