Product Details

SNP ID
rs138075306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37841792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACTGATACTTACCAATATATTT[A/G]TAGGCTTTTCCCAACTTAACCAGGT
Phenotype
MIM: 300302
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYNLT3 PubMed Links

Gene Details

Gene
DYNLT3
Gene Name
dynein light chain Tctex-type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006520.2 Intron NP_006511.1

View Full Product Details