Product Details

SNP ID
rs138172556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153786423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTTGGCGATGTTCTTATTGGC[A/G]ATACTCTTGCCGGTGTTCCTCGTAG
Phenotype
MIM: 300089 MIM: 300214 MIM: 300090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDH3G PubMed Links

Gene Details

Gene
IDH3G
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004135.3 1148 Silent Mutation ATC,ATT I317I NP_004126.1
NM_174869.2 1148 Silent Mutation ATC,ATT I317I NP_777358.1
Gene
PLXNB3
Gene Name
plexin B3
There are no transcripts associated with this gene.

Gene
SRPK3
Gene Name
SRSF protein kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170760.1 1148 Intron NP_001164231.1
NM_001170761.1 1148 Intron NP_001164232.1
NM_014370.3 1148 Intron NP_055185.2
Gene
SSR4
Gene Name
signal sequence receptor subunit 4
There are no transcripts associated with this gene.

View Full Product Details