Product Details
- SNP ID
-
rs139126908
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:155033372 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCTAGTTAGGTATAAAAGTAAAT[A/C]TTTTCTTATTTAACGTATTACTTCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FUNDC2
PubMed Links
Gene Details
- Gene
- FUNDC2
- Gene Name
- FUN14 domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023934.3 |
|
Intron |
|
|
NP_076423.2 |
View Full Product Details