Product Details

SNP ID
rs139449312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153767118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCCTGTAATCGACAGCCCTGA[C/G]TGCGTGCCCTTCCGTGACCCAGCCG
Phenotype
MIM: 300214
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLXNB3 PubMed Links

Gene Details

Gene
PLXNB3
Gene Name
plexin B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163257.1 465 Missense Mutation GAC,GAG D120E NP_001156729.1
NM_005393.2 465 Missense Mutation GAC,GAG D97E NP_005384.2

View Full Product Details