Product Details

SNP ID
rs140311832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47196470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATCTTCAGTTTGGAGACTCAG[A/G]TATGTTTAGAAACTAAGCCACAGCT
Phenotype
MIM: 300164 MIM: 300080 MIM: 314370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INE1 PubMed Links

Gene Details

Gene
INE1
Gene Name
inactivation escape 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RBM10
Gene Name
RNA binding motif protein 10
There are no transcripts associated with this gene.

Gene
UBA1
Gene Name
ubiquitin like modifier activating enzyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003334.3 2446 Intron NP_003325.2
NM_153280.2 2446 Intron NP_695012.1
XM_005272649.1 2446 Intron XP_005272706.1
XM_011543954.2 2446 Intron XP_011542256.1
XM_017029777.1 2446 Intron XP_016885266.1
XM_017029778.1 2446 Intron XP_016885267.1
XM_017029779.1 2446 UTR 5 XP_016885268.1
XM_017029780.1 2446 Intron XP_016885269.1
XM_017029781.1 2446 Intron XP_016885270.1

View Full Product Details