Product Details
- SNP ID
-
rs140496175
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:47223724 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACACGTGCTGCTCCTGGGGAAC[A/T]TCGTTCTGCACGGGGCCCACTCAGC
- Phenotype
-
MIM: 311550
MIM: 314370
MIM: 300050
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDK16
PubMed Links
Gene Details
- Gene
- CDK16
- Gene Name
- cyclin dependent kinase 16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001170460.1 |
1077 |
Missense Mutation |
CAT,CTT |
H130L |
NP_001163931.1 |
NM_006201.4 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
NP_006192.1 |
NM_033018.3 |
1077 |
Missense Mutation |
CAT,CTT |
H62L |
NP_148978.2 |
XM_011543920.2 |
1077 |
Missense Mutation |
CAT,CTT |
H130L |
XP_011542222.1 |
XM_011543921.1 |
1077 |
Missense Mutation |
CAT,CTT |
H103L |
XP_011542223.1 |
XM_011543922.1 |
1077 |
Missense Mutation |
CAT,CTT |
H99L |
XP_011542224.1 |
XM_011543923.1 |
1077 |
Missense Mutation |
CAT,CTT |
H62L |
XP_011542225.1 |
XM_011543924.1 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_011542226.1 |
XM_011543925.2 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_011542227.1 |
XM_011543926.1 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_011542228.1 |
XM_011543927.2 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_011542229.1 |
XM_011543928.1 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_011542230.1 |
XM_017029569.1 |
1077 |
Missense Mutation |
CAT,CTT |
H103L |
XP_016885058.1 |
XM_017029570.1 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_016885059.1 |
XM_017029571.1 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_016885060.1 |
XM_017029572.1 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_016885061.1 |
XM_017029573.1 |
1077 |
Missense Mutation |
CAT,CTT |
H56L |
XP_016885062.1 |
- Gene
- UBA1
- Gene Name
- ubiquitin like modifier activating enzyme 1
There are no transcripts associated with this gene.
- Gene
- USP11
- Gene Name
- ubiquitin specific peptidase 11
There are no transcripts associated with this gene.
View Full Product Details