Product Details

SNP ID
rs140966823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55452728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCTGGAAAGTGCTGGGGAAGTT[A/G]GGAATGCAGCCTGGACGTCAGCACA
Phenotype
MIM: 300548
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEH1 PubMed Links

Gene Details

Gene
MAGEH1
Gene Name
MAGE family member H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014061.4 640 Silent Mutation TTA,TTG L118L NP_054780.2
Gene
MIR4536-1
Gene Name
microRNA 4536-1
There are no transcripts associated with this gene.

Gene
MIR4536-2
Gene Name
microRNA 4536-2
There are no transcripts associated with this gene.

View Full Product Details