Product Details

SNP ID
rs141375838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101616411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTTCAGTCCAATCCCCATCCTC[A/G]GTCCAGGGCCGAGCCATAGTTTCAA
Phenotype
MIM: 300364
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMCX3 PubMed Links

Gene Details

Gene
ARMCX3
Gene Name
armadillo repeat containing, X-linked 3
There are no transcripts associated with this gene.

Gene
ARMCX6
Gene Name
armadillo repeat containing, X-linked 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009584.1 643 Silent Mutation ACC,ACT T70T NP_001009584.1
NM_001184768.1 643 Silent Mutation ACC,ACT T70T NP_001171697.1
NM_019007.3 643 Silent Mutation ACC,ACT T70T NP_061880.2
XM_011530976.2 643 Silent Mutation ACC,ACT T133T XP_011529278.1

View Full Product Details