Product Details

SNP ID
rs141450062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47198841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGTCCAAGAAACGTCGCGTGTC[C/T]GGGCCTGATCCAAAGCCGGGTTCTA
Phenotype
MIM: 300164 MIM: 314370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INE1 PubMed Links

Gene Details

Gene
INE1
Gene Name
inactivation escape 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
UBA1
Gene Name
ubiquitin like modifier activating enzyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003334.3 147 Silent Mutation TCC,TCT S13S NP_003325.2
NM_153280.2 147 Silent Mutation TCC,TCT S13S NP_695012.1
XM_005272649.1 147 Silent Mutation TCC,TCT S19S XP_005272706.1
XM_011543954.2 147 Silent Mutation TCC,TCT S27S XP_011542256.1
XM_017029777.1 147 Silent Mutation TCC,TCT S64S XP_016885266.1
XM_017029778.1 147 Silent Mutation TCC,TCT S41S XP_016885267.1
XM_017029779.1 147 Silent Mutation TCC,TCT S19S XP_016885268.1
XM_017029780.1 147 Silent Mutation TCC,TCT S13S XP_016885269.1
XM_017029781.1 147 Silent Mutation TCC,TCT S13S XP_016885270.1

View Full Product Details