Product Details

SNP ID
rs141847845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153781850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTGTGAGGCACCTCCCTACCC[C/T]ACTCCCAGCTCCCCTGGAGCTGCCT
Phenotype
MIM: 300089 MIM: 300214
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IDH3G PubMed Links

Gene Details

Gene
IDH3G
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) gamma
There are no transcripts associated with this gene.

Gene
PLXNB3
Gene Name
plexin B3
There are no transcripts associated with this gene.

Gene
SRPK3
Gene Name
SRSF protein kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170760.1 Intron NP_001164231.1
NM_001170761.1 Intron NP_001164232.1
NM_014370.3 Intron NP_055185.2

View Full Product Details