Product Details

SNP ID
rs142420567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103274357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGCCCTGCTTTTCCTGGTTTCC[C/T]TCATCTTCCAGTTGTCCCTCATCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCEAL5 PubMed Links

Gene Details

Gene
TCEAL5
Gene Name
transcription elongation factor A like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012979.2 499 Silent Mutation GAA,GAG E69E NP_001012997.1

View Full Product Details